Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| DENND2D - DENN domain containing 2D | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001271833.1 | Intron | NP_001258762.1 | ||||
| NM_024901.4 | Intron | NP_079177.2 | ||||
| XM_006710921.2 | Intron | XP_006710984.2 | ||||
| XM_006710922.1 | Intron | XP_006710985.1 | ||||
| XM_006710923.1 | Intron | XP_006710986.1 | ||||
| XM_006710924.1 | Intron | XP_006710987.1 | ||||
| XM_011542187.1 | Intron | XP_011540489.1 | ||||
| XM_011542189.1 | Intron | XP_011540491.1 | ||||
| XM_011542190.2 | Intron | XP_011540492.1 | ||||
| XM_017002386.1 | Intron | XP_016857875.1 | ||||
| XM_017002387.1 | Intron | XP_016857876.1 | ||||
| XM_017002388.1 | Intron | XP_016857877.1 | ||||
| XM_017002389.1 | Intron | XP_016857878.1 | ||||

DENND2D
Genomic Map