Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
PSG8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| PSG8 - pregnancy specific beta-1-glycoprotein 8 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001130167.1 | 600 | Missense Mutation | ACC,TCC | T,S 168 | NP_001123639.1 | |
| NM_001130168.1 | 600 | Missense Mutation | ACC,TCC | T,S 46 | NP_001123640.1 | |
| NM_182707.2 | 600 | Missense Mutation | ACC,TCC | T,S 168 | NP_874366.1 | |
| XM_011526986.1 | 600 | Intron | XP_011525288.1 | |||
| XM_011526987.2 | 600 | Intron | XP_011525289.1 | |||
| XM_011526988.2 | 600 | Intron | XP_011525290.1 | |||

PSG8
Genomic Map