Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||
Literature Links: |
C1orf105 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf105 - chromosome 1 open reading frame 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300760.1 | Intron | NP_001287689.1 | ||||
NM_139240.3 | Intron | NP_640333.3 | ||||
XM_005245608.2 | Intron | XP_005245665.1 | ||||
XM_005245610.3 | Intron | XP_005245667.1 | ||||
XM_006711630.2 | Intron | XP_006711693.1 | ||||
XM_011510152.2 | Intron | XP_011508454.1 | ||||
XM_011510153.2 | Intron | XP_011508455.1 | ||||
XM_011510154.2 | Intron | XP_011508456.1 | ||||
XM_011510155.2 | Intron | XP_011508457.1 | ||||
XM_011510156.2 | Intron | XP_011508458.1 | ||||
XM_017002808.1 | Intron | XP_016858297.1 |
PIGC - phosphatidylinositol glycan anchor biosynthesis class C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002642.3 | Intron | NP_002633.1 | ||||
NM_153747.1 | Intron | NP_714969.1 |
