Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GBA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GBA2 - glucosylceramidase beta 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020944.2 | 238 | Intron | NP_065995.1 | |||
| XM_005251526.4 | 238 | Intron | XP_005251583.1 | |||
| XM_006716809.3 | 238 | Intron | XP_006716872.1 | |||
| XM_011517973.2 | 238 | Intron | XP_011516275.1 | |||
| XM_017014937.1 | 238 | Intron | XP_016870426.1 | |||
| XM_017014938.1 | 238 | Intron | XP_016870427.1 | |||
| XM_017014939.1 | 238 | Intron | XP_016870428.1 | |||
| XM_017014940.1 | 238 | Intron | XP_016870429.1 | |||
| XM_017014941.1 | 238 | Intron | XP_016870430.1 | |||
| XM_017014942.1 | 238 | Intron | XP_016870431.1 | |||
| XM_017014943.1 | 238 | Intron | XP_016870432.1 | |||
| XM_017014944.1 | 238 | Intron | XP_016870433.1 | |||
| XM_017014945.1 | 238 | Intron | XP_016870434.1 | |||
| XM_017014946.1 | 238 | Intron | XP_016870435.1 | |||
| MSMP - microseminoprotein, prostate associated | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NPR2 - natriuretic peptide receptor 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| RGP1 - RGP1 homolog, RAB6A GEF complex partner 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001080496.2 | 238 | Silent Mutation | CTC,CTT | L,L 30 | NP_001073965.2 | |

GBA2
Genomic Map