Assay Details
| Species: | Human | ||||||||||||||||||||
| dbSNP Submissions: | NA | ||||||||||||||||||||
| Phenotype: |  MIM: 617007 | ||||||||||||||||||||
| Literature Links: | TRIM35 PubMed Links | ||||||||||||||||||||
| Allele Nomenclature: | |||||||||||||||||||||
| Minor Allele Frequency: | 
| 1000Genome | Applied Biosystems® | HapMap | 
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | 
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | 
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | 
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | 
| EUR - Not Available | ||
| AMR - Not Available | 
| TRIM35 - tripartite motif containing 35 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001304495.1 | Intron | NP_001291424.1 | ||||
| NM_171982.4 | Intron | NP_741983.2 | ||||
| XM_005273452.3 | Intron | XP_005273509.1 | ||||
| XM_011544453.1 | Intron | XP_011542755.1 | ||||
 Back To Top
 Back To Top
 TRIM35
TRIM35 Genomic Map
Genomic Map Back To Top
 Back To Top