Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
![]() ![]() |
||||||||||||||||||||
Literature Links: |
CHRNA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA2 - cholinergic receptor nicotinic alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000742.3 | 1360 | Frame Shift Delete | AAGATC,AGA | K,R I296 | NP_000733.2 | |
NM_001282455.1 | 1360 | Frame Shift Delete | AAGATC,AGA | K,R I281 | NP_001269384.1 | |
XM_005273397.2 | 1360 | Missense Mutation | AAGAGA,AAGATC | K,I R137K | XP_005273454.1 | |
XM_006716282.1 | 1360 | Missense Mutation | AAGAGA,AAGATC | K,I R296K | XP_006716345.1 | |
XM_011544388.1 | 1360 | Missense Mutation | AAGAGA,AAGATC | K,I R296K | XP_011542690.1 | |
XM_011544389.1 | 1360 | Missense Mutation | AAGAGA,AAGATC | K,I R98K | XP_011542691.1 | |
XM_017013003.1 | 1360 | Missense Mutation | AAGAGA,AAGATC | K,I R137K | XP_016868492.1 | |
XM_017013004.1 | 1360 | Missense Mutation | AAGAGA,AAGATC | K,I R98K | XP_016868493.1 |
PTK2B - protein tyrosine kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
