Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
![]() ![]() ![]() ![]() |
||||||||||||||||||||
Literature Links: |
DCTN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCTN2 - dynactin subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDIT3 - DNA damage inducible transcript 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195053.1 | Intron | NP_001181982.1 | ||||
NM_001195054.1 | Intron | NP_001181983.1 | ||||
NM_001195055.1 | Intron | NP_001181984.1 | ||||
NM_001195056.1 | Intron | NP_001181985.1 | ||||
NM_001195057.1 | Intron | NP_001181986.1 | ||||
NM_004083.5 | Intron | NP_004074.2 |
MARS - methionyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MBD6 - methyl-CpG binding domain protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052897.3 | Intron | NP_443129.3 | ||||
XM_005268617.1 | Intron | XP_005268674.1 | ||||
XM_005268618.1 | Intron | XP_005268675.1 | ||||
XM_005268620.2 | Intron | XP_005268677.1 | ||||
XM_006719217.1 | Intron | XP_006719280.1 | ||||
XM_006719218.1 | Intron | XP_006719281.1 | ||||
XM_011537851.2 | Intron | XP_011536153.1 |
MIR616 - microRNA 616 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6758 - microRNA 6758 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
