Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
HYPK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| HYPK - huntingtin interacting protein K | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MFAP1 - microfibrillar associated protein 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR1282 - microRNA 1282 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SERF2 - small EDRK-rich factor 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001018108.3 | 793 | Intron | NP_001018118.1 | |||
| NM_001199875.1 | 793 | Intron | NP_001186804.1 | |||
| NM_001199876.1 | 793 | Intron | NP_001186805.1 | |||
| NM_001199877.1 | 793 | Intron | NP_001186806.1 | |||
| NM_001199878.1 | 793 | Intron | NP_001186807.1 | |||
| SERF2-C15ORF63 - SERF2-C15orf63 readthrough | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SERINC4 - serine incorporator 4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001258031.1 | 793 | Missense Mutation | CGC,TGC | R,C 285 | NP_001244960.1 | |
| NM_001258032.1 | 793 | Missense Mutation | CGC,TGC | R,C 41 | NP_001244961.1 | |

HYPK
Genomic Map