Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
EIF2AK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| EIF2AK4 - eukaryotic translation initiation factor 2 alpha kinase 4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001013703.3 | Intron | NP_001013725.2 | ||||
| XM_005254392.2 | Intron | XP_005254449.1 | ||||
| XM_011521599.2 | Intron | XP_011519901.1 | ||||
| XM_011521600.2 | Intron | XP_011519902.1 | ||||
| XM_017022219.1 | Intron | XP_016877708.1 | ||||

EIF2AK4
Genomic Map