Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
BFAR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| BFAR - bifunctional apoptosis regulator | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_016561.2 | 2134 | UTR 3 | NP_057645.1 | |||
| XM_005255350.2 | 2134 | UTR 3 | XP_005255407.1 | |||
| XM_011522520.1 | 2134 | UTR 3 | XP_011520822.1 | |||
| PLA2G10 - phospholipase A2 group X | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

BFAR
Genomic Map