Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ACACA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ACACA - acetyl-CoA carboxylase alpha | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_198834.2 | 1172 | Intron | NP_942131.1 | |||
| NM_198836.2 | 1172 | Intron | NP_942133.1 | |||
| NM_198837.1 | 1172 | Intron | NP_942134.1 | |||
| NM_198838.1 | 1172 | Intron | NP_942135.1 | |||
| NM_198839.2 | 1172 | Intron | NP_942136.1 | |||
| XM_005257267.4 | 1172 | Intron | XP_005257324.1 | |||
| XM_006721853.1 | 1172 | Intron | XP_006721916.1 | |||
| XM_011524701.1 | 1172 | Intron | XP_011523003.1 | |||
| XM_011524703.1 | 1172 | Intron | XP_011523005.1 | |||
| XM_011524704.2 | 1172 | Intron | XP_011523006.1 | |||
| XM_017024553.1 | 1172 | Intron | XP_016880042.1 | |||
| XM_017024554.1 | 1172 | Intron | XP_016880043.1 | |||
| XM_017024555.1 | 1172 | Intron | XP_016880044.1 | |||
| C17orf78 - chromosome 17 open reading frame 78 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001321399.1 | 1172 | UTR 3 | NP_001308328.1 | |||
| NM_173625.4 | 1172 | UTR 3 | NP_775896.3 | |||
| XM_011524647.2 | 1172 | UTR 3 | XP_011522949.1 | |||
| XM_011524648.2 | 1172 | UTR 3 | XP_011522950.1 | |||
| XM_011524649.2 | 1172 | Intron | XP_011522951.1 | |||

ACACA
Genomic Map