Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ZNF234 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ZNF234 - zinc finger protein 234 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001144824.1 | Intron | NP_001138296.1 | ||||
| NM_006630.2 | Intron | NP_006621.1 | ||||
| XM_006722974.3 | Intron | XP_006723037.1 | ||||
| XM_017026149.1 | Intron | XP_016881638.1 | ||||
| XM_017026150.1 | Intron | XP_016881639.1 | ||||
| XM_017026151.1 | Intron | XP_016881640.1 | ||||
| XM_017026152.1 | Intron | XP_016881641.1 | ||||
| XM_017026153.1 | Intron | XP_016881642.1 | ||||

ZNF234
Genomic Map