Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
AP2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| AP2A1 - adaptor related protein complex 2 alpha 1 subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_014203.2 | Intron | NP_055018.2 | ||||
| NM_130787.2 | Intron | NP_570603.2 | ||||
| XM_011526556.2 | Intron | XP_011524858.1 | ||||
| XM_011526557.2 | Intron | XP_011524859.1 | ||||
| MIR6799 - microRNA 6799 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

AP2A1
Genomic Map