Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
SLC7A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| SLC7A9 - solute carrier family 7 member 9 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001126335.1 | Intron | NP_001119807.1 | ||||
| NM_001243036.1 | Intron | NP_001229965.1 | ||||
| NM_014270.4 | Intron | NP_055085.1 | ||||
| XM_006722992.1 | Intron | XP_006723055.1 | ||||
| XM_011526402.2 | Intron | XP_011524704.1 | ||||
| XM_017026230.1 | Intron | XP_016881719.1 | ||||

SLC7A9
Genomic Map