Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| C21orf58 - chromosome 21 open reading frame 58 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001006114.2 | Intron | NP_001006114.1 | ||||
| NM_001314022.1 | Intron | NP_001300951.1 | ||||
| NM_001314023.1 | Intron | NP_001300952.1 | ||||
| NM_001314024.1 | Intron | NP_001300953.1 | ||||
| NM_001314025.1 | Intron | NP_001300954.1 | ||||
| NM_001314026.1 | Intron | NP_001300955.1 | ||||
| NM_058181.2 | Intron | NP_478061.1 | ||||
| XM_005261156.3 | Intron | XP_005261213.2 | ||||
| XM_011529629.2 | Intron | XP_011527931.1 | ||||
| XM_011529630.2 | Intron | XP_011527932.1 | ||||
| XM_011529631.2 | Intron | XP_011527933.1 | ||||
| XM_011529633.2 | Intron | XP_011527935.1 | ||||
| XM_011529634.2 | Intron | XP_011527936.2 | ||||
| XM_011529635.2 | Intron | XP_011527937.1 | ||||
| XM_017028394.1 | Intron | XP_016883883.1 | ||||
| XM_017028395.1 | Intron | XP_016883884.1 | ||||
| XM_017028396.1 | Intron | XP_016883885.1 | ||||

C21orf58
Genomic Map