Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| PCNT - pericentrin | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001315529.1 | 472 | Intron | NP_001302458.1 | |||
| NM_006031.5 | 472 | Intron | NP_006022.3 | |||
| XM_005261124.4 | 472 | Intron | XP_005261181.1 | |||
| XM_011529594.2 | 472 | Intron | XP_011527896.1 | |||
| XM_017028362.1 | 472 | Intron | XP_016883851.1 | |||
| XM_017028363.1 | 472 | Intron | XP_016883852.1 | |||
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

C21orf58
Genomic Map