Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| C21orf58 - chromosome 21 open reading frame 58 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001286462.1 | Intron | NP_001273391.1 | ||||
| NM_001286463.1 | Intron | NP_001273392.1 | ||||
| NM_001286476.1 | Intron | NP_001273405.1 | ||||
| NM_001286477.1 | Intron | NP_001273406.1 | ||||
| NM_058180.4 | Intron | NP_478060.2 | ||||
| XM_005261148.4 | Intron | XP_005261205.1 | ||||
| XM_006724018.3 | Intron | XP_006724081.1 | ||||
| XM_011529619.2 | Intron | XP_011527921.1 | ||||
| XM_011529620.2 | Intron | XP_011527922.1 | ||||
| XM_011529621.2 | Intron | XP_011527923.1 | ||||
| XM_011529623.2 | Intron | XP_011527925.1 | ||||
| XM_011529624.2 | Intron | XP_011527926.1 | ||||
| XM_011529625.2 | Intron | XP_011527927.1 | ||||
| XM_011529626.2 | Intron | XP_011527928.1 | ||||
| XM_011529627.2 | Intron | XP_011527929.1 | ||||
| XM_011529628.1 | Intron | XP_011527930.1 | ||||
| XM_017028388.1 | Intron | XP_016883877.1 | ||||
| XM_017028389.1 | Intron | XP_016883878.1 | ||||
| XM_017028390.1 | Intron | XP_016883879.1 | ||||
| XM_017028391.1 | Intron | XP_016883880.1 | ||||
| XM_017028392.1 | Intron | XP_016883881.1 | ||||
| XM_017028393.1 | Intron | XP_016883882.1 | ||||
| PCNT - pericentrin | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001006114.2 | Intron | NP_001006114.1 | ||||
| NM_001314022.1 | Intron | NP_001300951.1 | ||||
| NM_001314023.1 | Intron | NP_001300952.1 | ||||
| NM_001314024.1 | Intron | NP_001300953.1 | ||||
| NM_001314025.1 | Intron | NP_001300954.1 | ||||
| NM_001314026.1 | Intron | NP_001300955.1 | ||||
| NM_058181.2 | Intron | NP_478061.1 | ||||
| XM_005261156.3 | Intron | XP_005261213.2 | ||||
| XM_011529629.2 | Intron | XP_011527931.1 | ||||
| XM_011529630.2 | Intron | XP_011527932.1 | ||||
| XM_011529631.2 | Intron | XP_011527933.1 | ||||
| XM_011529633.2 | Intron | XP_011527935.1 | ||||
| XM_011529634.2 | Intron | XP_011527936.2 | ||||
| XM_011529635.2 | Intron | XP_011527937.1 | ||||
| XM_017028394.1 | Intron | XP_016883883.1 | ||||
| XM_017028395.1 | Intron | XP_016883884.1 | ||||
| XM_017028396.1 | Intron | XP_016883885.1 | ||||

C21orf58
Genomic Map