Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GALR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GALR3 - galanin receptor 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| GCAT - glycine C-acetyltransferase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001171690.1 | Intron | NP_001165161.1 | ||||
| NM_014291.3 | Intron | NP_055106.1 | ||||
| XM_005261409.3 | Intron | XP_005261466.1 | ||||
| XM_005261410.3 | Intron | XP_005261467.1 | ||||
| XM_005261411.1 | Intron | XP_005261468.1 | ||||
| XM_011530023.1 | Intron | XP_011528325.1 | ||||
| XM_017028674.1 | Intron | XP_016884163.1 | ||||
| XM_017028675.1 | Intron | XP_016884164.1 | ||||
| XM_017028676.1 | Intron | XP_016884165.1 | ||||
| XM_017028677.1 | Intron | XP_016884166.1 | ||||
| H1F0 - H1 histone family member 0 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

GALR3
Genomic Map