Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
RBBP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| RBBP4 - RB binding protein 4, chromatin remodeling factor | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ZBTB8OS - zinc finger and BTB domain containing 8 opposite strand | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001308135.1 | Intron | NP_001295064.1 | ||||
| NM_001308136.1 | Intron | NP_001295065.1 | ||||
| NM_001308137.1 | Intron | NP_001295066.1 | ||||
| NM_001308138.1 | Intron | NP_001295067.1 | ||||
| NM_001308139.1 | Intron | NP_001295068.1 | ||||
| NM_001308140.1 | Intron | NP_001295069.1 | ||||
| NM_001308141.1 | Intron | NP_001295070.1 | ||||
| NM_178547.3 | Intron | NP_848642.1 | ||||
| XM_011541327.2 | Intron | XP_011539629.1 | ||||
| XM_011541328.1 | Intron | XP_011539630.1 | ||||
| XM_011541329.2 | Intron | XP_011539631.1 | ||||
| XM_011541330.2 | Intron | XP_011539632.1 | ||||
| XM_011541334.2 | Intron | XP_011539636.1 | ||||
| XM_011541335.2 | Intron | XP_011539637.1 | ||||
| XM_011541337.2 | Intron | XP_011539639.1 | ||||
| XM_011541338.2 | Intron | XP_011539640.1 | ||||
| XM_017001134.1 | Intron | XP_016856623.1 | ||||
| XM_017001135.1 | Intron | XP_016856624.1 | ||||
| XM_017001136.1 | Intron | XP_016856625.1 | ||||
| XM_017001137.1 | Intron | XP_016856626.1 | ||||
| XM_017001138.1 | Intron | XP_016856627.1 | ||||

RBBP4
Genomic Map