Assay Details
| Species: | Human | ||||||||||||||||||||
| dbSNP Submissions: | 5 submissions | ||||||||||||||||||||
| Phenotype: |  MIM: 615183  MIM: 176982 | ||||||||||||||||||||
| Literature Links: | FAAP20 PubMed Links | ||||||||||||||||||||
| Allele Nomenclature: | |||||||||||||||||||||
| Minor Allele Frequency: | 
| 1000Genome | Applied Biosystems® | HapMap | 
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | 
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | 
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | 
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | 
| EUR - Not Available | ||
| AMR - Not Available | 
| FAAP20 - Fanconi anemia core complex associated protein 20 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC100506504 - uncharacterized LOC100506504 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| XM_011542503.2 | 875 | Missense Mutation | CCG,CTG | P,L 83 | XP_011540805.1 | |
 Back To Top
 Back To Top
 FAAP20
FAAP20 Genomic Map
Genomic Map Back To Top
 Back To Top