Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||
Literature Links: |
TRAF3IP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAF3IP1 - TRAF3 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139490.1 | Intron | NP_001132962.1 | ||||
NM_015650.3 | Intron | NP_056465.2 | ||||
XM_006712414.2 | Intron | XP_006712477.1 | ||||
XM_011510944.2 | Intron | XP_011509246.1 | ||||
XM_011510945.2 | Intron | XP_011509247.1 | ||||
XM_011510946.2 | Intron | XP_011509248.1 | ||||
XM_011510947.2 | Intron | XP_011509249.1 | ||||
XM_011510948.2 | Intron | XP_011509250.1 | ||||
XM_011510950.2 | Intron | XP_011509252.1 | ||||
XM_017003789.1 | Intron | XP_016859278.1 |
