Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
NCKAP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| NCKAP5 - NCK associated protein 5 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_207363.2 | Intron | NP_997246.2 | ||||
| NM_207481.3 | Intron | NP_997364.3 | ||||
| XM_005263659.3 | Intron | XP_005263716.1 | ||||
| XM_005263660.3 | Intron | XP_005263717.1 | ||||
| XM_011511097.2 | Intron | XP_011509399.1 | ||||
| XM_011511098.2 | Intron | XP_011509400.1 | ||||
| XM_011511099.2 | Intron | XP_011509401.1 | ||||
| XM_011511100.2 | Intron | XP_011509402.1 | ||||
| XM_011511101.1 | Intron | XP_011509403.1 | ||||
| XM_011511102.2 | Intron | XP_011509404.1 | ||||
| XM_011511103.2 | Intron | XP_011509405.1 | ||||
| XM_011511104.1 | Intron | XP_011509406.1 | ||||
| XM_011511105.2 | Intron | XP_011509407.1 | ||||
| XM_017003974.1 | Intron | XP_016859463.1 | ||||
| XM_017003975.1 | Intron | XP_016859464.1 | ||||
| XM_017003976.1 | Intron | XP_016859465.1 | ||||
| XM_017003977.1 | Intron | XP_016859466.1 | ||||
| XM_017003978.1 | Intron | XP_016859467.1 | ||||
| XM_017003979.1 | Intron | XP_016859468.1 | ||||
| XM_017003980.1 | Intron | XP_016859469.1 | ||||

NCKAP5
Genomic Map