Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ANKRD37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ANKRD37 - ankyrin repeat domain 37 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_181726.3 | 2207 | Missense Mutation | ATG,GTG | M,V 139 | NP_859077.1 | |
| XM_017008176.1 | 2207 | Missense Mutation | ATG,GTG | M,V 139 | XP_016863665.1 | |
| C4orf47 - chromosome 4 open reading frame 47 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LRP2BP - LRP2 binding protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| UFSP2 - UFM1 specific peptidase 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_018359.3 | 2207 | UTR 3 | NP_060829.2 | |||

ANKRD37
Genomic Map