Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GRID2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GRID2 - glutamate ionotropic receptor delta type subunit 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001286838.1 | Intron | NP_001273767.1 | ||||
| NM_001510.3 | Intron | NP_001501.2 | ||||
| XM_011531893.2 | Intron | XP_011530195.2 | ||||
| XM_011531894.2 | Intron | XP_011530196.1 | ||||
| XM_011531895.2 | Intron | XP_011530197.1 | ||||
| XM_017008118.1 | Intron | XP_016863607.1 | ||||
| XM_017008119.1 | Intron | XP_016863608.1 | ||||
| XM_017008120.1 | Intron | XP_016863609.1 | ||||
| XM_017008121.1 | Intron | XP_016863610.1 | ||||
| XM_017008122.1 | Intron | XP_016863611.1 | ||||
| XM_017008123.1 | Intron | XP_016863612.1 | ||||
| XM_017008124.1 | Intron | XP_016863613.1 | ||||
| XM_017008125.1 | Intron | XP_016863614.1 | ||||
| XM_017008126.1 | Intron | XP_016863615.1 | ||||
| XM_017008127.1 | Intron | XP_016863616.1 | ||||

GRID2
Genomic Map