Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LRP2BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| LRP2BP - LRP2 binding protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_018409.3 | 5164 | UTR 3 | NP_060879.2 | |||
| XM_005263125.4 | 5164 | Intron | XP_005263182.1 | |||
| XM_006714260.3 | 5164 | UTR 3 | XP_006714323.1 | |||
| XM_006714265.3 | 5164 | Intron | XP_006714328.1 | |||
| XM_011532110.2 | 5164 | Intron | XP_011530412.1 | |||
| XM_011532111.2 | 5164 | Intron | XP_011530413.1 | |||
| XM_017008410.1 | 5164 | UTR 3 | XP_016863899.1 | |||
| XM_017008411.1 | 5164 | Intron | XP_016863900.1 | |||
| SNX25 - sorting nexin 25 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001317781.1 | 5164 | Intron | NP_001304710.1 | |||
| XM_005263267.4 | 5164 | Intron | XP_005263324.1 | |||
| XM_011532302.2 | 5164 | Intron | XP_011530604.1 | |||
| XM_011532303.2 | 5164 | Intron | XP_011530605.1 | |||
| XM_011532304.2 | 5164 | Intron | XP_011530606.1 | |||
| XM_011532305.2 | 5164 | Intron | XP_011530607.1 | |||
| XM_011532306.2 | 5164 | Intron | XP_011530608.1 | |||
| XM_011532307.2 | 5164 | Intron | XP_011530609.1 | |||
| XM_011532308.2 | 5164 | Intron | XP_011530610.1 | |||
| XM_017008680.1 | 5164 | Intron | XP_016864169.1 | |||
| XM_017008681.1 | 5164 | Intron | XP_016864170.1 | |||
| XM_017008682.1 | 5164 | Intron | XP_016864171.1 | |||
| XM_017008683.1 | 5164 | Intron | XP_016864172.1 | |||
| XM_017008684.1 | 5164 | Intron | XP_016864173.1 | |||
| XM_017008685.1 | 5164 | Intron | XP_016864174.1 | |||
| XM_017008686.1 | 5164 | Intron | XP_016864175.1 | |||
| XM_017008687.1 | 5164 | Intron | XP_016864176.1 | |||

LRP2BP
Genomic Map