Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ANKRD37 - ankyrin repeat domain 37 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LRP2BP - LRP2 binding protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_018409.3 | Intron | NP_060879.2 | ||||
| XM_005263125.4 | Intron | XP_005263182.1 | ||||
| XM_006714260.3 | Intron | XP_006714323.1 | ||||
| XM_006714265.3 | Intron | XP_006714328.1 | ||||
| XM_011532110.2 | Intron | XP_011530412.1 | ||||
| XM_011532111.2 | Intron | XP_011530413.1 | ||||
| XM_017008410.1 | Intron | XP_016863899.1 | ||||
| XM_017008411.1 | Intron | XP_016863900.1 | ||||

ANKRD37
Genomic Map