Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||
Literature Links: |
PDE4D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE4D - phosphodiesterase 4D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104631.1 | Intron | NP_001098101.1 | ||||
NM_001165899.1 | Intron | NP_001159371.1 | ||||
NM_001197218.1 | Intron | NP_001184147.1 | ||||
NM_001197219.1 | Intron | NP_001184148.1 | ||||
NM_001197220.1 | Intron | NP_001184149.1 | ||||
NM_001197221.1 | Intron | NP_001184150.1 | ||||
NM_001197222.1 | Intron | NP_001184151.1 | ||||
NM_001197223.1 | Intron | NP_001184152.1 | ||||
NM_006203.4 | Intron | NP_006194.2 | ||||
XM_005248537.3 | Intron | XP_005248594.1 | ||||
XM_005248538.4 | Intron | XP_005248595.1 | ||||
XM_011543469.1 | Intron | XP_011541771.1 | ||||
XM_011543470.2 | Intron | XP_011541772.1 | ||||
XM_011543471.1 | Intron | XP_011541773.1 | ||||
XM_011543473.1 | Intron | XP_011541775.1 | ||||
XM_011543477.2 | Intron | XP_011541779.1 | ||||
XM_017009565.1 | Intron | XP_016865054.1 | ||||
XM_017009566.1 | Intron | XP_016865055.1 | ||||
XM_017009567.1 | Intron | XP_016865056.1 | ||||
XM_017009568.1 | Intron | XP_016865057.1 | ||||
XM_017009569.1 | Intron | XP_016865058.1 | ||||
XM_017009570.1 | Intron | XP_016865059.1 | ||||
XM_017009571.1 | Intron | XP_016865060.1 |
