Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||
Literature Links: |
MIR4804 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4804 - microRNA 4804 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNPO1 - transportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002270.3 | Intron | NP_002261.3 | ||||
NM_153188.2 | Intron | NP_694858.1 | ||||
XM_005248500.2 | Intron | XP_005248557.1 | ||||
XM_005248501.1 | Intron | XP_005248558.1 | ||||
XM_017009458.1 | Intron | XP_016864947.1 |
