Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
PARK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| PARK2 - parkin RBR E3 ubiquitin protein ligase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004562.2 | Intron | NP_004553.2 | ||||
| NM_013987.2 | Intron | NP_054642.2 | ||||
| NM_013988.2 | Intron | NP_054643.2 | ||||
| XM_011535863.1 | Intron | XP_011534165.1 | ||||
| XM_017010908.1 | Intron | XP_016866397.1 | ||||
| XM_017010909.1 | Intron | XP_016866398.1 | ||||

PARK2
Genomic Map