Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ITGB2 - integrin subunit beta 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000211.4 | Intron | NP_000202.3 | ||||
| NM_001127491.2 | Intron | NP_001120963.2 | ||||
| NM_001303238.1 | Intron | NP_001290167.1 | ||||
| XM_006724001.1 | Intron | XP_006724064.1 | ||||
| XM_017028341.1 | Intron | XP_016883830.1 | ||||
| ITGB2-AS1 - ITGB2 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LINC01547 - long intergenic non-protein coding RNA 1547 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

ITGB2
Genomic Map