Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2A7P1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| CYP2A7P1 - cytochrome P450 family 2 subfamily A member 7 pseudogene 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| CYP2B6 - cytochrome P450 family 2 subfamily B member 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000767.4 | Intron | NP_000758.1 | ||||
| XM_005258569.4 | Intron | XP_005258626.1 | ||||
| XM_006723050.3 | Intron | XP_006723113.1 | ||||
| XM_011526546.2 | Intron | XP_011524848.1 | ||||
| XM_011526547.2 | Intron | XP_011524849.1 | ||||
| XM_011526548.2 | Intron | XP_011524850.1 | ||||
| XM_011526549.2 | Intron | XP_011524851.1 | ||||
| XM_011526550.2 | Intron | XP_011524852.1 | ||||
More Information
Set Membership: |
|

CYP2A7P1
Genomic Map