Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS9D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| VPS9D1 - VPS9 domain containing 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004913.2 | 2307 | Silent Mutation | ATC,ATT | I,I 104 | NP_004904.2 | |
| XM_005256329.4 | 2307 | Silent Mutation | ATC,ATT | I,I 104 | XP_005256386.1 | |
| XM_005256330.4 | 2307 | Silent Mutation | ATC,ATT | I,I 34 | XP_005256387.1 | |
| XM_005256331.2 | 2307 | Intron | XP_005256388.1 | |||
| XM_006721350.2 | 2307 | Silent Mutation | ATC,ATT | I,I 104 | XP_006721413.1 | |
| XM_011523476.2 | 2307 | Silent Mutation | ATC,ATT | I,I 103 | XP_011521778.1 | |
| XM_011523477.2 | 2307 | Silent Mutation | ATC,ATT | I,I 70 | XP_011521779.1 | |
| XM_011523478.2 | 2307 | Silent Mutation | ATC,ATT | I,I 69 | XP_011521780.1 | |
| XM_011523479.2 | 2307 | Silent Mutation | ATC,ATT | I,I 104 | XP_011521781.1 | |
| XM_011523480.2 | 2307 | Silent Mutation | ATC,ATT | I,I 34 | XP_011521782.1 | |
| VPS9D1-AS1 - VPS9D1 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ZNF276 - zinc finger protein 276 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
More Information
Set Membership: |
|

VPS9D1
Genomic Map