Assay Details
        Species: | 
      Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
        dbSNP Submissions: | 
      
        
          
          
            33 submissions
          
        
        
         | 
    ||||||||||||||||||||||||||||||||||||||||||||||||||
        Phenotype: | 
      
          
            
              
                 | 
      ||||||||||||||||||||||||||||||||||||||||||||||||||
        Literature Links: | 
      RIMS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
          Allele Nomenclature: | 
        |||||||||||||||||||||||||||||||||||||||||||||||||||
          Minor Allele Frequency: | 
        
        
          
            
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
						
						Global
						
						
					
					 
					
  | 
			
				Caucasian - Not Available | CEPH (CEU)
					 
					
  | 
				
		||||||
						
						EAS
					
					 
					
  | 
			
				African American - Not Available | YRI (Yoruba)
					 
					
  | 
				
		||||||
						
						SAS
					
					 
					
  | 
			
				Chinese - Not Available | JPT (Japanese)
					 
					
  | 
				
		||||||
						
						AFR
					
					 
					
  | 
			
				Japanese - Not Available | CHB (Han Chinese)
					 
					
  | 
				
		||||||
						
						EUR
					
					 
					
  | 
			
				||||||||
						
						AMR
					
					 
					
  | 
			
				
| RIMS3 - regulating synaptic membrane exocytosis 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_014747.2 | Intron | NP_055562.2 | ||||
| XM_011542479.2 | Intron | XP_011540781.1 | ||||
More Information
Set Membership: | 
		   
					
						
							 | 
				

RIMS3
Genomic Map