Assay Details
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| MOK - MOK protein kinase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001272011.1 | Intron | NP_001258940.1 | ||||
| NM_014226.2 | Intron | NP_055041.1 | ||||
| XM_006720225.2 | Intron | XP_006720288.1 | ||||
| XM_006720226.2 | Intron | XP_006720289.1 | ||||
| XM_011537052.1 | Intron | XP_011535354.1 | ||||
| XM_011537053.2 | Intron | XP_011535355.1 | ||||
| XM_017021549.1 | Intron | XP_016877038.1 | ||||
| XM_017021550.1 | Intron | XP_016877039.1 | ||||
| XM_017021551.1 | Intron | XP_016877040.1 | ||||
| XM_017021552.1 | Intron | XP_016877041.1 | ||||
| XM_017021553.1 | Intron | XP_016877042.1 | ||||
| XM_017021554.1 | Intron | XP_016877043.1 | ||||
| XM_017021555.1 | Intron | XP_016877044.1 | ||||
| XM_017021556.1 | Intron | XP_016877045.1 | ||||
| XM_017021557.1 | Intron | XP_016877046.1 | ||||
| XM_017021558.1 | Intron | XP_016877047.1 | ||||
| XM_017021559.1 | Intron | XP_016877048.1 | ||||
More Information
Set Membership: |
|

MOK
Genomic Map