Assay Details
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STARD13 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| STARD13 - StAR related lipid transfer domain containing 13 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001243466.1 | Intron | NP_001230395.1 | ||||
| NM_001243474.1 | Intron | NP_001230403.1 | ||||
| NM_001243476.2 | Intron | NP_001230405.1 | ||||
| NM_052851.2 | Intron | NP_443083.1 | ||||
| NM_178006.3 | Intron | NP_821074.1 | ||||
| NM_178007.2 | Intron | NP_821075.1 | ||||
| XM_011535299.2 | Intron | XP_011533601.1 | ||||
| XM_017020834.1 | Intron | XP_016876323.1 | ||||
| XM_017020835.1 | Intron | XP_016876324.1 | ||||
More Information
Set Membership: |
|

STARD13
Genomic Map