Assay Details
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||
Literature Links: |
PEX5L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
| EUR - Not Available | |||||
| AMR - Not Available |

PEX5L
Genomic Map