Assay Details
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf47 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| C4orf47 - chromosome 4 open reading frame 47 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| CCDC110 - coiled-coil domain containing 110 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001145411.1 | 1213 | Missense Mutation | ATG,CTG | M,L 345 | NP_001138883.1 | |
| NM_152775.3 | 1213 | Missense Mutation | ATG,CTG | M,L 382 | NP_689988.1 | |
| XM_005262891.3 | 1213 | Missense Mutation | ATG,CTG | M,L 382 | XP_005262948.1 | |
| XM_006714172.2 | 1213 | Missense Mutation | ATG,CTG | M,L 382 | XP_006714235.1 | |
| XM_011531827.2 | 1213 | Missense Mutation | ATG,CTG | M,L 382 | XP_011530129.1 | |
| XM_011531828.2 | 1213 | Missense Mutation | ATG,CTG | M,L 362 | XP_011530130.1 | |
| XM_011531829.2 | 1213 | Missense Mutation | ATG,CTG | M,L 312 | XP_011530131.1 | |
| XM_017007987.1 | 1213 | Missense Mutation | ATG,CTG | M,L 312 | XP_016863476.1 | |
| XM_017007988.1 | 1213 | Missense Mutation | ATG,CTG | M,L 312 | XP_016863477.1 | |
More Information
Set Membership: |
|

C4orf47
Genomic Map