Assay Details
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSBP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| SSBP2 - single stranded DNA binding protein 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001256732.1 | Intron | NP_001243661.1 | ||||
| NM_001256733.1 | Intron | NP_001243662.1 | ||||
| NM_001256734.1 | Intron | NP_001243663.1 | ||||
| NM_001256735.1 | Intron | NP_001243664.1 | ||||
| NM_001256736.1 | Intron | NP_001243665.1 | ||||
| NM_012446.3 | Intron | NP_036578.2 | ||||
| XM_011543297.2 | Intron | XP_011541599.1 | ||||
| XM_017009304.1 | Intron | XP_016864793.1 | ||||
| XM_017009305.1 | Intron | XP_016864794.1 | ||||
| XM_017009306.1 | Intron | XP_016864795.1 | ||||
| XM_017009307.1 | Intron | XP_016864796.1 | ||||
| XM_017009308.1 | Intron | XP_016864797.1 | ||||
| XM_017009309.1 | Intron | XP_016864798.1 | ||||
| XM_017009310.1 | Intron | XP_016864799.1 | ||||
More Information
Set Membership: |
|

SSBP2
Genomic Map