Assay Details
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||
Literature Links: |
SFMBT1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
| EUR - Not Available | |||||
| AMR - Not Available |
| SFMBT1 - Scm-like with four mbt domains 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_016329.3 | 7832 | Intron | NP_057413.2 | |||
| XM_005265221.3 | 7832 | UTR 3 | XP_005265278.1 | |||
| XM_006713203.3 | 7832 | UTR 3 | XP_006713266.1 | |||
| XM_006713204.3 | 7832 | UTR 3 | XP_006713267.1 | |||
| XM_011533824.2 | 7832 | UTR 3 | XP_011532126.1 | |||
| XM_011533825.2 | 7832 | UTR 3 | XP_011532127.1 | |||
| TMEM110 - transmembrane protein 110 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TMEM110-MUSTN1 - TMEM110-MUSTN1 readthrough | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

SFMBT1
Genomic Map