Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf49 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| C11orf49 - chromosome 11 open reading frame 49 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001003676.2 | Intron | NP_001003676.1 | ||||
| NM_001003677.2 | Intron | NP_001003677.1 | ||||
| NM_001003678.2 | Intron | NP_001003678.1 | ||||
| NM_001278222.1 | Intron | NP_001265151.1 | ||||
| NM_024113.4 | Intron | NP_077018.1 | ||||
| XM_006718315.1 | Intron | XP_006718378.1 | ||||
| XM_011520364.2 | Intron | XP_011518666.1 | ||||
| XM_011520365.2 | Intron | XP_011518667.1 | ||||
| XM_017018276.1 | Intron | XP_016873765.1 | ||||
| XM_017018277.1 | Intron | XP_016873766.1 | ||||
| XM_017018278.1 | Intron | XP_016873767.1 | ||||
| XM_017018279.1 | Intron | XP_016873768.1 | ||||
| XM_017018280.1 | Intron | XP_016873769.1 | ||||
| XM_017018281.1 | Intron | XP_016873770.1 | ||||
| XM_017018282.1 | Intron | XP_016873771.1 | ||||

C11orf49
Genomic Map