Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
DMPK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| DMPK - dystrophia myotonica protein kinase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001081560.2 | 3303 | Intron | NP_001075029.1 | |||
| NM_001081562.2 | 3303 | Intron | NP_001075031.1 | |||
| NM_001081563.2 | 3303 | Intron | NP_001075032.1 | |||
| NM_001288764.1 | 3303 | Intron | NP_001275693.1 | |||
| NM_001288765.1 | 3303 | Intron | NP_001275694.1 | |||
| NM_001288766.1 | 3303 | Intron | NP_001275695.1 | |||
| NM_004409.4 | 3303 | Intron | NP_004400.4 | |||
| DMWD - dystrophia myotonica, WD repeat containing | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004943.1 | 3303 | UTR 3 | NP_004934.1 | |||

DMPK
Genomic Map