Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| PLPP6 - phospholipid phosphatase 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_203453.3 | 959 | Missense Mutation | AGT,CGT | S,R 293 | NP_982278.3 | |
| SPATA6L - spermatogenesis associated 6 like | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001039395.3 | 959 | Intron | NP_001034484.3 | |||
| XM_006716795.2 | 959 | Intron | XP_006716858.1 | |||
| XM_006716798.3 | 959 | Intron | XP_006716861.1 | |||
| XM_006716799.3 | 959 | Intron | XP_006716862.1 | |||
| XM_006716800.2 | 959 | Intron | XP_006716863.1 | |||
| XM_006716801.2 | 959 | Intron | XP_006716864.1 | |||
| XM_011517952.1 | 959 | Intron | XP_011516254.1 | |||
| XM_011517954.1 | 959 | Intron | XP_011516256.1 | |||
| XM_011517956.1 | 959 | Intron | XP_011516258.1 | |||
| XM_011517957.1 | 959 | Intron | XP_011516259.1 | |||
| XM_011517958.1 | 959 | Intron | XP_011516260.1 | |||
| XM_017014881.1 | 959 | Intron | XP_016870370.1 | |||
| XM_017014882.1 | 959 | Intron | XP_016870371.1 | |||
| XM_017014883.1 | 959 | Intron | XP_016870372.1 | |||
| XM_017014884.1 | 959 | Intron | XP_016870373.1 | |||
| XM_017014885.1 | 959 | Intron | XP_016870374.1 | |||
| XM_017014886.1 | 959 | Intron | XP_016870375.1 | |||
| XM_017014887.1 | 959 | Intron | XP_016870376.1 | |||

PLPP6
Genomic Map