Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100996664 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| LOC100996664 - uncharacterized LOC100996664 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| RAD51B - RAD51 paralog B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001321809.1 | Intron | NP_001308738.1 | ||||
| NM_001321810.1 | Intron | NP_001308739.1 | ||||
| NM_001321812.1 | Intron | NP_001308741.1 | ||||
| NM_001321814.1 | Intron | NP_001308743.1 | ||||
| NM_001321815.1 | Intron | NP_001308744.1 | ||||
| NM_001321817.1 | Intron | NP_001308746.1 | ||||
| NM_001321818.1 | Intron | NP_001308747.1 | ||||
| NM_001321819.1 | Intron | NP_001308748.1 | ||||
| NM_001321821.1 | Intron | NP_001308750.1 | ||||
| NM_002877.5 | Intron | NP_002868.1 | ||||
| NM_133509.3 | Intron | NP_598193.2 | ||||
| NM_133510.3 | Intron | NP_598194.1 | ||||
| XM_011537050.2 | Intron | XP_011535352.1 | ||||
| XM_011537051.2 | Intron | XP_011535353.1 | ||||
| XM_017021545.1 | Intron | XP_016877034.1 | ||||
| XM_017021546.1 | Intron | XP_016877035.1 | ||||
| XM_017021547.1 | Intron | XP_016877036.1 | ||||
| XM_017021548.1 | Intron | XP_016877037.1 | ||||

LOC100996664
Genomic Map