Assay Details
| Species: | Human | ||||||||||||||||||||||||||||||||||||||
| dbSNP Submissions: | NA | ||||||||||||||||||||||||||||||||||||||
| Phenotype: |  MIM: 616150 | ||||||||||||||||||||||||||||||||||||||
| Literature Links: | LOC340074 PubMed Links | ||||||||||||||||||||||||||||||||||||||
| Allele Nomenclature: | |||||||||||||||||||||||||||||||||||||||
| Minor Allele Frequency: | 
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
| Global 
 | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
| EAS 
 | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS 
 | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR 
 | Japanese - Not Available | JPT (Japanese) - Not Available | |||
| EUR 
 | |||||
| AMR 
 | 
| LOC340074 - uncharacterized LOC340074 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SLC25A48 - solute carrier family 25 member 48 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_145282.4 | Intron | NP_660325.4 | ||||
| XM_005271903.1 | Intron | XP_005271960.1 | ||||
| XM_006714544.1 | Intron | XP_006714607.1 | ||||
| XM_006714546.1 | Intron | XP_006714609.1 | ||||
| XM_006714550.1 | Intron | XP_006714613.1 | ||||
| XM_006714552.2 | Intron | XP_006714615.1 | ||||
| XM_011543196.1 | Intron | XP_011541498.1 | ||||
| XM_011543199.1 | Intron | XP_011541501.1 | ||||
| XM_017009086.1 | Intron | XP_016864575.1 | ||||
| XM_017009087.1 | Intron | XP_016864576.1 | ||||
| XM_017009088.1 | Intron | XP_016864577.1 | ||||
| XM_017009089.1 | Intron | XP_016864578.1 | ||||
| XM_017009090.1 | Intron | XP_016864579.1 | ||||
| XM_017009091.1 | Intron | XP_016864580.1 | ||||
| XM_017009092.1 | Intron | XP_016864581.1 | ||||
 Back To Top
 Back To Top
 LOC340074
LOC340074 Genomic Map
Genomic Map Back To Top
 Back To Top