Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITM2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ITM2C - integral membrane protein 2C | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001012514.2 | 292 | Intron | NP_001012532.1 | |||
| NM_001012516.2 | 292 | Silent Mutation | GCC,GCT | A,A 75 | NP_001012534.1 | |
| NM_001287240.1 | 292 | Silent Mutation | GCC,GCT | A,A 13 | NP_001274169.1 | |
| NM_001287241.1 | 292 | Silent Mutation | GCC,GCT | A,A 75 | NP_001274170.1 | |
| NM_030926.5 | 292 | Silent Mutation | GCC,GCT | A,A 75 | NP_112188.1 | |

ITM2C
Genomic Map