Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| C21orf58 - chromosome 21 open reading frame 58 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001006114.2 | 1024 | Intron | NP_001006114.1 | |||
| NM_001314022.1 | 1024 | UTR 3 | NP_001300951.1 | |||
| NM_001314023.1 | 1024 | UTR 3 | NP_001300952.1 | |||
| NM_001314024.1 | 1024 | UTR 3 | NP_001300953.1 | |||
| NM_001314025.1 | 1024 | UTR 3 | NP_001300954.1 | |||
| NM_001314026.1 | 1024 | Intron | NP_001300955.1 | |||
| NM_058181.2 | 1024 | UTR 3 | NP_478061.1 | |||
| XM_005261156.3 | 1024 | UTR 3 | XP_005261213.2 | |||
| XM_011529629.2 | 1024 | Intron | XP_011527931.1 | |||
| XM_011529630.2 | 1024 | Intron | XP_011527932.1 | |||
| XM_011529631.2 | 1024 | Intron | XP_011527933.1 | |||
| XM_011529633.2 | 1024 | Intron | XP_011527935.1 | |||
| XM_011529634.2 | 1024 | Intron | XP_011527936.2 | |||
| XM_011529635.2 | 1024 | Intron | XP_011527937.1 | |||
| XM_017028394.1 | 1024 | Intron | XP_016883883.1 | |||
| XM_017028395.1 | 1024 | Intron | XP_016883884.1 | |||
| XM_017028396.1 | 1024 | Intron | XP_016883885.1 | |||
More Information
Set Membership: |
|

C21orf58
Genomic Map