Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| FANCC - Fanconi anemia complementation group C | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000136.2 | Intron | NP_000127.2 | ||||
| NM_001243743.1 | Intron | NP_001230672.1 | ||||
| NM_001243744.1 | Intron | NP_001230673.1 | ||||
| XM_005251802.3 | Intron | XP_005251859.1 | ||||
| XM_006717001.2 | Intron | XP_006717064.1 | ||||
| XM_006717002.3 | Intron | XP_006717065.1 | ||||
| XM_006717004.3 | Intron | XP_006717067.1 | ||||
| XM_011518365.2 | Intron | XP_011516667.1 | ||||
| XM_011518366.2 | Intron | XP_011516668.1 | ||||
| XM_011518367.2 | Intron | XP_011516669.1 | ||||
| XM_017014452.1 | Intron | XP_016869941.1 | ||||
| XM_017014453.1 | Intron | XP_016869942.1 | ||||
| XM_017014454.1 | Intron | XP_016869943.1 | ||||
More Information
Set Membership: |
|

C9orf3
Genomic Map