Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP2L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| BAIAP2L1 - BAI1 associated protein 2 like 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_018842.4 | Intron | NP_061330.2 | ||||
| BRI3 - brain protein I3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001159491.1 | Intron | NP_001152963.1 | ||||
| NM_015379.4 | Intron | NP_056194.1 | ||||
| XM_017011931.1 | Intron | XP_016867420.1 | ||||
| XM_017011932.1 | Intron | XP_016867421.1 | ||||
| XM_017011933.1 | Intron | XP_016867422.1 | ||||
| XM_017011934.1 | Intron | XP_016867423.1 | ||||
| XM_017011935.1 | Intron | XP_016867424.1 | ||||
| XM_017011936.1 | Intron | XP_016867425.1 | ||||
More Information
Set Membership: |
|

BAIAP2L1
Genomic Map