Assay Details
| Species: | Human | ||||||||||||||||||||||||||||||||||||||
| dbSNP Submissions: | NA | ||||||||||||||||||||||||||||||||||||||
| Phenotype: |  MIM: 601015  MIM: 609999 | ||||||||||||||||||||||||||||||||||||||
| Literature Links: | MIR4709 PubMed Links | ||||||||||||||||||||||||||||||||||||||
| Allele Nomenclature: | |||||||||||||||||||||||||||||||||||||||
| Minor Allele Frequency: | 
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
| Global 
 | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
| EAS 
 | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS 
 | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR 
 | Japanese - Not Available | JPT (Japanese) - Not Available | |||
| EUR 
 | |||||
| AMR 
 | 
| MIR4709 - microRNA 4709 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NPC2 - NPC intracellular cholesterol transporter 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_006432.3 | Intron | NP_006423.1 | ||||
| XM_017020930.1 | Intron | XP_016876419.1 | ||||
| SYNDIG1L - synapse differentiation inducing 1 like | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
 Back To Top
 Back To Top
 MIR4709
MIR4709 Genomic Map
Genomic Map Back To Top
 Back To Top