Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GRK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GRK6 - G protein-coupled receptor kinase 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001004105.2 | Intron | NP_001004105.1 | ||||
| NM_001004106.2 | Intron | NP_001004106.1 | ||||
| NM_002082.3 | Intron | NP_002073.2 | ||||
| XM_006714858.3 | Intron | XP_006714921.1 | ||||
| XM_006714859.2 | Intron | XP_006714922.1 | ||||
| XM_011534537.2 | Intron | XP_011532839.1 | ||||
| XM_011534538.1 | Intron | XP_011532840.1 | ||||
| PRR7 - proline rich 7 (synaptic) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PRR7-AS1 - PRR7 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

GRK6
Genomic Map